Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62P Mouse anti-Human, FITC, Clone: AK-4, BD

Catalog No. BDB555523 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555523 100 Tests
BDB561922 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB555523 Supplier BD Biosciences Supplier No. 555523
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The AK-4 monoclonal antibody specifically binds to CD62P. CD62P is a 140 kDa type I transmembrane glycoprotein that is also known as P-Selectin, Platelet activation-dependent granule membrane protein (PADGEM), or GMP-140. P-Selectin is stored in the α -granules of platelets and the Weibel-Palade bodies of endothelial cells, and is rapidly transported to the plasma membrane upon activation. P-Selectin is thought to mediate the initial adhesive interactions of neutrophils and monocytes with endothelium in inflammatory responses, and of activated platelets to neutrophils and monocytes in hemostasis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD62P
Applications Flow Cytometry
Classification Monoclonal
Clone AK-4
Conjugate FITC
Description P-Selectin; GMP-140; PADGEM
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.