Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam CD63 Exosome Capture Beads, Human, Liquid, 25 Test
Description
- Target: CD63
- Application: Exosome capture
- Reactive species: Human
- Form: Liquid
- Preservative: 0.1% Sodium azide
Specifications
Specifications
| Reactivity | Human |
| Target Molecule | CD63 |
| Format | Beads |
| No. of Reactions | 25 Tests |
| Buffer | 0.1% sodium azide |
| Form | Liquid |
| Cross Reactivity | Human |
| Formulation | 0.1% Sodium azide |
| Quantity | 25 Tests |
| For Use With (Application) | Exosome capture |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?