Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD63 Rabbit, anti-Human, Unconjugated, MP Biomedicals™

Catalog No. MP08L100021 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08L100021 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP08L100021 Supplier MP Biomedicals, Inc Supplier No. 08L100021
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD63 Rabbit Monoclonal Antibody has been experimentally validated in WB and IHC assays.

  • The protein encoded by this gene is a member of the transmembrane 4 superfamily; they mediate signal transduction events that play a role in the regulation of cell development, activation, growth, and motility
  • Offers high specificity and potency against CD36 antigen in immunological and biochemical assays and consistency between lots
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD63
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution WB: 1:500 to 1:2,000; IHC: 1:50 to 1:200
Formulation In PBS buffer with 0.02% sodium azide, 0.05% BSA, 50% glycerol, pH7.3 to 7.4. This antibody can be stored at 2 to 8°C for one month without detectable loss of activity. This antibody is stable for twelve months from the date of receipt when stored at -20 to -80*deg;C. Avoid repeated freeze-thaw cycles.
Gene Alias CD63; LAMP-3; ME491; MLA1; OMA81H; TSPAN30; CD63 antigen; CD63
Host Species Rabbit
Immunogen Full-length human CD63 protein as an immunogen
Purification Method Affinity Purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage -20°C
Form Liquid
Isotype IgG
Show More Show Less

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.