Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD65 Monoclonal Antibody (VIM8), FITC, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. p-7068666
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-245-238 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-245-238 Supplier Invitrogen™ Supplier No. 11065942
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This VIM8 antibody reacts with the desialylated asialoganglioside of human CD65. This type 2 fucosylated carbohydrate antigen is expressed on the surface of all myeloid cells during development. Moreover, CD65 is expressed highly on granulocytes and weakly on monocytes in peripheral blood. CD65 has been reported to be a ligand for CD62E (E-Selectin). However, the sialylated form of CD65 (designated CD65s) can be used as a marker for acute myeloid leukemia (AML) as its expression increases as CD34 decreases. Finally, CD65 is important for extravascular infiltration of AML cells. Applications Reported: This VIM8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This VIM8 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.

This type 2 fucosylated carbohydrate antigen is expressed on the surface of all myeloid cells during development. Moreover, CD65 is expressed highly on granulocytes and weakly on monocytes in peripheral blood. CD65 has been reported to be a ligand for CD62E (E-Selectin). However, the sialylated form of CD65 (designated CD65s) can be used as a marker for acute myeloid leukemia (AML) as its expression increases as CD34 decreases. Finally, CD65 is important for extravascular infiltration of AML cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD65
Applications Flow Cytometry
Classification Monoclonal
Clone VIM8
Concentration 5 μL/Test
Conjugate FITC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Alias Fucoganglioside Type 2; Fucoganglioside Type II
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.