Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD66 Monoclonal Antibody (CD66a-B1.1), APC, eBioscience™
GREENER_CHOICE

Catalog No. 50245657
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-245-657 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-245-657 Supplier Invitrogen™ Supplier No. 17066842
  • $398.00
    $298.65 / Each of 1

    Save $99.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The CD66a-B1.1 monoclonal antibody reacts with human CD66a, c, d, and e, which are also known as CEACAM1, CEACAM6, CEACAM3 and CEA, respectively. CD66 is expressed on endothelial and epithelial cells, as well as on neutrophils and monocytes. Myeloma and cancer cells also express CD66. Moreover, expression of this receptor can be induced on T cells, B cells, and CD16-negative NK cells. CD66 exists as multiple splice variants that differ in the length of their cytoplasmic domain, which contains an immunoreceptor tyrosine-based inhibitory motif (ITIM) that mediates the inhibitory functions of this cell surface protein on T and NK cells. CD66 is involved in cell adhesion and growth, neutrophil activation, and signaling. Finally, studies have shown that the CD66 variants interact with each other homo- and heterotypically. Applications Reported: This CD66a-B1.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This CD66a-B1.1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

The CD66 molecules are 180-200 kDa glycoproteins of carcinoembryonic antigen family. They are present on all blood granulocytes and some tissue macrophages, but are absent from other hematopoietic cells. The expression of CD66 increases significantly on granulocytes upon their activation. CD66a (BGP1, CEACAM1) and CD66d (CGM1, CEACAM3), as well as CEACAM2 and 4, are transmembrane proteins, whereas CD66c (CEAL, NCA, CEACAM6) and CD66e (CEA, CEACAM5), as well as CEACAM7 and 8, are anchored to the plasma membrane by C-terminal glycosylphosphatidylinositol (GPI) lipid moiety.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD66
Applications Flow Cytometry
Classification Monoclonal
Clone CD66a-B1.1
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CEACAM4
Gene Accession No. O75871, P06731, P13688, P40198, P40199
Gene Alias antigen CD66; BGP; BGP1; BGP-1; BGPI; Biliary glycoprotein 1; Carcinoembryonic antigen; Carcinoembryonic antigen CGM1; carcinoembryonic antigen CGM7; carcinoembryonic antigen gene family member 1; carcinoembryonic antigen gene family member 7; carcinoembryonic antigen related cell adhesion molecule 1; carcinoembryonic antigen related cell adhesion molecule 3; carcinoembryonic antigen related cell adhesion molecule 4; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein); carcinoembryonic antigen-related cell adhesion molecule 3; Carcinoembryonic antigen-related cell adhesion molecule 4; carcinoembryonic antigen-related cell adhesion molecule 4-sv1; carcinoembryonic antigen-related cell adhesion molecule 4-sv2; carcinoembryonic antigen-related cell adhesion molecule 5; Carcinoembryonic antigen-related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen); CD66a; CD66a antigen; CD66c; CD66d; CD66d antigen; CD66e; CEA; CEACAM1; CEACAM-1; CEACAM3; CEACAM-3; CEACAM4; CEACAM5; CEACAM-5; CEACAM6; CEACAM-6; CEAL; CGM1; CGM7; CGM7_HUMAN; Cluster of Differentiation 66c; meconium antigen 100; NCA; Non-specific crossreacting antigen; nonspecific cross-reacting antigen; Nonspecific cross-reacting antigen (NCA); non-specific cross-reacting antigen W236; nonspecific cross-reacting antigen W236; normal cross-reacting antigen; pan CEACAM; W264; W282
Gene Symbols CEACAM1, CEACAM3, CEACAM4, CEACAM5, CEACAM6
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1048, 1084, 1089, 4680, 634
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.