Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD68 Recombinant Mouse Monoclonal Antibody (PDM0-13)
GREENER_CHOICE

Catalog No. FEMA550651
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550651 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550651 Supplier Invitrogen™ Supplier No. MA550651
Only null left
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Positive control: Human spleen tissue, human diffuse large B-cell lymphoma tissue, human tonsil tissue, human liver tissue, THP-1, human non-small cell lung cancer, human cervical cancer. Predicted band size: 37 kDa Subcellular Location: Cell membrane Endosome membrane, Lysosome membrane.

CD68 (Macrosialin) is a 110 kDa integral membrane glycoprotein predominantly expressed on the intracellular lysomsomes of monocytes and macrophages and to a lesser extent by dendritic cells and peripheral blood granulocytes. Also, CD68 could play a role in phagocytic activities of tissue macrophages, both in intracellular lysosomal metabolism and extracellular cell-cell and cell-pathogen interactions. CD68 is expressed by interdigitating reticulum cells in tonsil and some histiocytic lymphoma or histiocytosis, acute myeloid leukemia (AML), and granulocytic sarcoma. Elevated expression of CD68 has been demonstrated on CD34+ cells in various human malignancies, including several Acute Myeloid Leukemia studies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD68
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Immunocytochemistry
Classification Recombinant Monoclonal
Clone PDM0-13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Cd68
Gene Accession No. P34810
Gene Alias CD68; CD68 antigen; CD68 molecule; GP110; Lamp4; macrophage antigen CD68; macrosialin; Scard1; scavenger receptor class D, member 1; similar to CD68 antigen
Gene Symbols Cd68
Host Species Mouse
Immunogen Recombinant protein within Human CD68 aa 1-354.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 968
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.