Learn More
Genscript Corporation CD69/CLEC2C His Cynomolgus
Supplier:  Genscript Corporation Z04125500
Encompass_Preferred
CLEC2C (CD69) is a membrane-bound type II C-lectin receptor and acts as a costimulatory molecule for T cell activation and proliferation It is involved in lymphocyte proliferation and functions as a signal transmitting receptor in lymphocytes natural killer (NK) cells and platelets CLEC2C is a disulfide-linked homodimer protein with two differentially glycosylated subunits
Catalog No. 50-000-58463
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.