Learn More
Invitrogen™ CD69 Monoclonal Antibody (FN50), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Description
Also called AIM or activation inducer molecule, CD69 is a gp28/34 disulfide bonded homodimer with a Mr of 60kDa under non-reducing conditions. CD69 contains one or two N linked oligosacaride. The CD69 molecule is also present on activated platelets. In normal peripheral blood a variable percentage of cells express the CD69 antigen. It is involved in lymphocyte signal transduction.
Specifications
Specifications
| Antigen | CD69 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | FN50 |
| Concentration | 5 μL/Test |
| Conjugate | PerCP-eFluor 710 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | CD69 |
| Gene Accession No. | Q07108 |
| Gene Alias | 5830438K24Rik; Activation inducer molecule; activation inducer molecule (AIM/CD69); AI452015; AIM; BL-AC/P26; CD69; CD69 antigen; CD69 antigen (p60, early T-cell activation antigen); CD69 molecule; CLEC2C; C-type lectin domain family 2 member C; C-type lectin domain family 2, member C; EA1; early activation antigen CD69; early lymphocyte activation antigen; early T-cell activation antigen p60; GP32/28; leukocyte surface antigen Leu-23; MLR-3; VEA; Very Early Activation Antigen |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.