Learn More
Invitrogen™ CD7 Monoclonal Antibody (4H9), PE, eBioscience™

Description
Description: This 4H9 monoclonal antibody reacts with human CD7 (also known as Leu-9 or gp40), a 40-kDa transmembrane glycoprotein belonging to the immunoglobulin superfamily. This molecule is expressed on peripheral blood T lymphocytes and natural killer cells, as well as on lymphoid and myeloid lineage precursors during adult hematopoiesis. Capable of providing a costimulatory signal and associating with CD3 and CD45, CD7 is involved in T cell activation. CD7 has also been detected on numerous T cell-derived leukemias and lymphomias. In fact, because CD7 expression is greater on CD4+ T cells in classical Hodgkin's lymphoma than in benign reactive lymph adenopathy, this molecule may be used as a diagnostic for Hodgkin's lymphoma. Blocking assays demonstrate that this clone recognizes the same epitope as clone 124-1D1. Applications Reported: This 4H9 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 4H9 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: μ8 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 4H9 |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | CD7 |
| Gene Accession No. | P09564 |
| Gene Alias | Cd7; CD7 antigen; CD7 antigen (p41); Cd7 molecule; GP40; LEU-9; p41 protein; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen Leu-9; Tp40; TP41 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.