Learn More
Invitrogen™ CD7 Monoclonal Antibody (eBio124-1D1 (124-1D1)), Brilliant Ultra Violet™ 737, eBioscience™, Invitrogen™

Description
CD7 Monoclonal antibody specifically detects CD7 in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | CD7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | eBio124-1D1 (124-1D1) |
| Concentration | 5 μL/Test |
| Conjugate | Brilliant Ultraviolet 737 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | CD7 |
| Gene Accession No. | P09564 |
| Gene Alias | Cd7; CD7 antigen; CD7 antigen (p41); Cd7 molecule; GP40; LEU-9; p41 protein; T-cell antigen CD7; T-cell leukemia antigen; T-cell surface antigen Leu-9; Tp40; TP41 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.