Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD70 Recombinant Rabbit Monoclonal Antibody (7F4S5)
GREENER_CHOICE

Catalog No. PIMA542758
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIMA542758 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIMA542758

Supplier: Invitrogen™ MA542758

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: Raji, 293T. Immunogen sequence: QRFAQAQQQL PLESLGWDVA ELQLNHTGPQ QDPRLYWQGG PALGRSFLHG PELDKGQLRI HRDGIYMVHI QVTLAICSST TASRHHPTTL AVGICSPASR SISLLRLSFH QGCTIASQRL TPLARGDTLC TNLTGTLLPS RNTDETFFGV QWVRP.

The protein encoded by this gene is a cytokine that belongs to the tumor necrosis factor (TNF) ligand family. This cytokine is a ligand for TNFRSF27/CD27. It is a surface antigen on activated, but not on resting, T and B lymphocytes. It induces proliferation of costimulated T cells, enhances the generation of cytolytic T cells, and contributes to T cell activation. This cytokine is also reported to play a role in regulating B-cell activation, cytotoxic function of natural killer cells, and immunoglobulin synthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD70
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 7F4S5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Cd70
Gene Accession No. P32970
Gene Alias CD27 ligand; Cd27l; CD27-L; CD27LG; CD70; CD70 antigen; Cd70 molecule; Ki-24 antigen; surface antigen CD70; Tnfsf7; TNLG8A; tumor necrosis factor (ligand) superfamily, member 7; tumor necrosis factor ligand 8A; tumor necrosis factor ligand superfamily member 7
Gene Symbols Cd70
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 970
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.