Learn More
Invitrogen™ CD72 Monoclonal Antibody (10.1.D2), PE

Description
CD72 Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | CD72 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 10.1.D2 |
| Concentration | 0.1 mg/mL |
| Conjugate | PE |
| Formulation | PBS with <0.1% sodium azide; pH 7.5 |
| Gene | CD72 |
| Gene Accession No. | P21855 |
| Gene Alias | B-cell differentiation antigen CD72; CD72; CD72 antigen; Cd72 molecule; CD72b; CD72c; Ly-19; Ly32; Ly-32; LYB2; Lyb-2; Ly-m19; lymphocyte antigen 32; RP11-331F9.9 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.