Learn More
CD79a/mb-1 (B-Cell Marker) Ab-1 Mouse Monoclonal Antibody, Epredia™
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunofluorescence experiments.
$691.00 - $706.00
Specifications
Antigen | CD79a/mb-1 (B-Cell Marker) Ab-1 |
---|---|
Clone | HM47/A9 |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Catalog Number | Mfr. No. | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
MS357B1
|
Epredia
MS357B1 |
Concentrated | Biotin | 500 μL |
Each for $706.00
|
|
|||
MS357P1
|
Epredia
MS357P1 |
Concentrated | Unconjugated | 500 μL |
Each for $691.00
|
|
|||
MS357B0
|
Epredia
MS357B0 |
Concentrated | Biotin | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS357B
|
Epredia
MS357B |
Concentrated | Biotin | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS357P0
|
Epredia
MS357P0 |
Concentrated | Unconjugated | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS357P1ABX
|
Epredia
MS357P1ABX |
Concentrated | Unconjugated | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS357PABX
|
Epredia
MS357PABX |
Concentrated | Unconjugated | 200 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS357P
|
Epredia
MS357P |
Concentrated | Unconjugated | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS357R1
|
Epredia
MS357R1 |
Prediluted | Unconjugated | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS357R7
|
Epredia
MS357R7 |
Prediluted | Unconjugated | 7 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines.Host Species: Mouse
Clone: HM47/A9
Isotype: IgG1/κ
Species Reactivity: Human, Monkey, Horse, Cow, Pig, Rabbit, Guinea Pig, Rat, Mouse and Chicken (Plasma cells). Does not react with opossum.
Epitope: GTYQDVGSLNIADVQ
Immunogen: A synthetic peptide consisting of GTYQDVGSLNIADVQ of human CD79a protein
Molecular Weight: 47 kDa
Positive Control: Raji cells, Lymph node or tonsil
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
CD79a/mb-1 (B-Cell Marker) Ab-1 | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Mouse | |
B-Cell | |
A synthetic peptide consisting of GTYQDVGSLNIADVQ of human CD79a protein | |
Primary |
HM47/A9 | |
Monoclonal | |
RUO | |
Bovine, Chicken, Equine, Guinea Pig, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat | |
IgG2a κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.