Learn More
Invitrogen™ CD79a Monoclonal Antibody (HM57), FITC

Description
This antibody The antibody HM57 interacts with intracellular domain of CD79a (Ig alpha), a 40-45 kDa subunit of B cell antigen-specific receptor (BCR) and its early developmental forms. Recommended dilution is for 100 μL of whole blood or 1x10^6 cells in suspension.
Specifications
Specifications
| Antigen | CD79a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HM57 |
| Concentration | 55 μg/mL |
| Conjugate | FITC |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| Gene | CD79A |
| Gene Accession No. | P11911, P11912, P40293 |
| Gene Alias | B-cell antigen receptor; B-cell antigen receptor complex-associated protein alpha chain; B-cell antigen receptor complex-associated protein alpha chain-like; CD79; CD79a; CD79A antigen (immunoglobulin-associated alpha); CD79a molecule; CD79a molecule, immunoglobulin-associated alpha; CD79A protein; CD79-alpha protein; EGK_10665; Ig alpha; IGA; Igalpha; ig-alpha; immunoglobulin-associated alpha; LOC100913063; Ly54; Ly-54; MB1; MB-1; MB-1 membrane glycoprotein; Membrane-bound immunoglobulin-associated protein; surface IgM-associated protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.