Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD8/CD28 Antibody Cocktail, FITC, PE
GREENER_CHOICE

Catalog No. p-7171292
Change view
Click to view available options
Quantity:
100 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-885 100 Tests
01-670-886 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-670-885 Supplier Invitrogen™ Supplier No. MA112166
Only null left
Add to Cart
Add to Cart

Antibody Cocktail

This antibody is a monoclonal mixture of FITC-conjugated CD8 (clone 17D8, isotype IgG1) and PE-conjugated CD28 (clone B-23, isotype IgG1).

CD8 molecule is composed of two chains termed alpha and beta. CD8 is found on a T cell subset of normal cytotoxic / suppressor cells which make up approximately 20 to 35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer cells, 80% of thymocytes, on a subpopulation of 30% of peripheral blood null cells and 15 to 30% of bone marrow cells. Human CD28 antigen is a 44 kDa disulfide linked homodimeric T cell specific surface glycoprotein. It is a cell adhesion molecule of the immunoglobulin superfamily which is constitutively expressed on most peripheral blood T lymphocytes (approximately 95% of CD4+ cells and 50% of CD8+ cells). Mature thymocytes exhibit higher levels of CD28 than the immature cells. Activation of T cells results in enhanced CD28 expression. T cell activation requires two combined signals provided by antigen presenting cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8/CD28
Applications Flow Cytometry
Classification Cocktail
Clone 17D8, B-23, B-23
Conjugate FITC/PE
Formulation PBS with 0.2% carrier protein and 0.08% sodium azide
Gene CD28
Gene Accession No. P01732, P10747, P10966
Gene Alias CD28; CD28 antigen; CD28 molecule; CD8; CD8 antigen, alpha polypeptide (p32); CD8 antigen, beta polypeptide 1 (p37); CD8A; CD8a molecule; CD8alpha; CD8b; CD8b molecule; CD8B1; CD8beta; LEU2; Leu-2; Leu2 T-lymphocyte antigen; LY3; LYT3; MAL; MGC138290; OKT8 T-cell antigen; p32; P37; T cell co-receptor; T lymphocyte surface glycoprotein beta chain; T8 T-cell antigen; T-cell antigen Leu2; T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-cell-specific surface glycoprotein CD28; T-lymphocyte differentiation antigen T8/Leu-2; Tp44
Gene Symbols CD28, CD8A, CD8B
Host Species Mouse
Immunogen human peripheral blood T lymphocytes and HPB-ALL T-cell line.
Purification Method Protein A/G
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 925, 926, 940
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.