Learn More
Invitrogen™ CD8/CD45RA Antibody Cocktail, FITC, PE
Antibody Cocktail
Supplier:  Invitrogen™ MA112229
Catalog No. 01-670-937
Only null left
Add to Cart
Description
Description
This antibody is a monoclonal mixture of FITC-conjugated CD8 (clone 17D8, isotype IgG1) and PE-conjugated CD45RA (clone BHKG-2, isotype IgG1).
CD8 molecule is composed of two chains termed alpha and beta. CD8 is found on a T cell subset of normal cytotoxic / suppressor cells which make up approximately 20 to 35 % of human peripheral blood lymphocytes. The CD8 antigen is also detected on natural killer cells, 80% of thymocytes, on a subpopulation of 30% of peripheral blood null cells and 15 to 30% of bone marrow cells. CD45 Leukocyte Common Antigen (LCA) belongs to the family of at least four isoforms of membrane glycoproteins (220, 205, 190, 180 kDa) expressed on hematopoietic cell lines but absent on non-hematopoietic cell lines, normal and malignant, non-hematopoietic tissues. The intracellular portion of these molecules have protein phosphatase activity and are involved in regulation of transmembrane signals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD8/CD45RA
Cocktail
FITC/PE
PTPRC
B220; CD45; CD45 antigen; CD45R; CD8; CD8 antigen, alpha polypeptide (p32); CD8 antigen, beta polypeptide 1 (p37); CD8a; CD8a molecule; CD8alpha; CD8B; CD8b molecule; CD8B1; CD8beta; GP180; LCA; L-CA; LEU2; Leu-2; Leu2 T-lymphocyte antigen; Leukocyte common antigen; LY3; LY5; LYT3; MAL; OKT8 T-cell antigen; p32; P37; protein tyrosine phosphatase, receptor type C; protein tyrosine phosphatase, receptor type, C; protein tyrosine phosphatase, receptor type, c polypeptide; PTPRC; Receptor-type tyrosine-protein phosphatase C; T cell co-receptor; T lymphocyte surface glycoprotein beta chain; T200; T200 glycoprotein; T200 leukocyte common antigen; T8 T-cell antigen; T-cell antigen Leu2; T-cell surface glycoprotein CD8 alpha chain; T-cell surface glycoprotein CD8 beta chain; T-lymphocyte differentiation antigen T8/Leu-2
Mouse
Protein A/G
RUO
5788, 925, 926
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
17D8, BHKG-2, BHKG-2
PBS with 0.2% carrier protein and 0.08% sodium azide
P01732, P08575, P10966
CD8A, CD8B, PTPRC
human peripheral blood monocytes from a rheumatoid arthritis patient and human peripheral blood T lymphocytes.
100 Tests
Primary
Human
Antibody
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.