Learn More
CAPRICO BIOTECHNOLOGIES INC CD8 iFluor 488 Antibody, 4006115
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4006115
Encompass_Preferred
The Caprico CD8 iFluor 488 Antibody is a mouse monoclonal antibody that specifically binds to human CD8. It is conjugated to iFluor 488, a bright green-fluorescent dye. This antibody can be used for flow cytometry, immunofluorescence microscopy, and other fluorescent detection methods. It recognizes the 32-34 kDa CD8 antigen, which is expressed on cytotoxic T cells, thymocytes, and NK cells. This antibody is supplied as a liquid in PBS with 0.1% sodium azide and 1% BSA. The concentration is 0.5 mg/mL. Recommended dilutions are 1:100-1:400 for flow cytometry and 1:50-1:200 for immunofluorescence. This antibody is validated for human samples.
Catalog No. 50-227-9270
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.