Learn More
CAPRICO BIOTECHNOLOGIES INC CD8 PE Antibody, 4006025
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4006025
Encompass_Preferred
The Caprico CD8 PE Antibody is a mouse monoclonal antibody that specifically binds to human CD8. It is conjugated to R-phycoerythrin (PE) and is intended for flow cytometry applications. This antibody recognizes the 32-34 kDa alpha chain of CD8. CD8 is expressed on cytotoxic T cells and acts as a co-receptor in T cell activation. This antibody can be used to identify cytotoxic T lymphocytes and is recommended for staining peripheral blood mononuclear cells. The concentration is 0.2 mg/mL with 0.09% sodium azide as a preservative. The clone is UCHT-4.
Catalog No. 50-227-9276
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.