Learn More
CAPRICO BIOTECHNOLOGIES INC CD8 PerCP Antibody, 4006035
Supplier:  CAPRICO BIOTECHNOLOGIES INC 4006035
Encompass_Preferred
The Caprico CD8 PerCP Antibody is a mouse monoclonal antibody that specifically binds to human CD8. It is conjugated to PerCP and is suitable for flow cytometry, immunohistochemistry, and immunofluorescence. This antibody recognizes the 32-34 kDa alpha chain of human CD8, also known as CD8a. CD8 is expressed on cytotoxic T cells and acts as a co-receptor for MHC class I restricted TCRs. This antibody can be used to identify cytotoxic T cells and is recommended for use in multicolor flow cytometry panels. The recommended dilution for flow cytometry is 5-25 μl per test and the optimal dilution should be determined by titration.
Catalog No. 50-227-9273
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.