Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD81 Monoclonal Antibody (1D6-CD81), APC, eBioscience™
GREENER_CHOICE

Catalog No. 501123064
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-3064 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-3064 Supplier Invitrogen™ Supplier No. 17081942
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: This 1D6-CD81 monoclonal antibody reacts with human CD81, which is also known as Target of Anti-Proliferative Ab-1 (TAPA-1). A member of the tetraspanin superfamily of transmembrane proteins, CD81 is widely expressed on immune cells such as B, T, and NK cells, monocytes, and eosinophils. Studies suggest that the highest expression of CD81 can be found on germinal center B cells. This protein can also be detected on non-Hodgkin lymphomas and diffuse large B-cell lymphomas. On B cells, CD81 exists in a complex with CD19, CD21, and Leu13. CD81 plays a role in segregating the CD19/CD21-B cell receptor complexes to lipid rafts to activate signal transduction. Finally, CD81 is a receptor for hepatitis C virus. The 1D6 monoclonal antibody has been reported to induce adhesion and reduce cell proliferation. Applications Reported: This 1D6-CD81 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1D6-CD81 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

CD81 (TAPA-1, target of anti-proliferative antibody-1) is a member of the tetraspanin family, is expressed on virtually all nucleated cells, but above all on germinal center B cells. CD81 forms complexes with other tetraspanin proteins, integrins, coreceptors, MHC class I and II molecules, and influences adhesion, morphology, activation, proliferation and differentiation of B, T cells. In muscles, CD81 promotes cell fusion and myotube maintenance. CD81 has been also identified as a receptor for the hepatitis C virus. Like members of the tetraspanin family that include CD9, CD37, CD53, CD63, and CD82, CD81 is a cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. CD81 is a cell surface glycoprotein that is known to complex with integrins. CD81 appears to promote muscle cell fusion and support myotube maintenance. CD81 associates with CD19, CD21, Leu 13, and integrins on cell membrane and acts as a receptor for the envelope protein E2 of chronic hepatitis C virus. Antibodies to CD81 have anti-proliferative effects on different lymphoid cell lines, particularly those derived from large cell lymphomas. CD81 is also localized in the tumor-suppressor gene region and is a candidate gene for malignancies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD81
Applications Flow Cytometry
Classification Monoclonal
Clone 1D6-CD81
Concentration 5 μL/Test
Conjugate APC
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene CD81
Gene Accession No. P60033
Gene Alias 26 kDa cell surface protein TAPA-1; CD 81 antigen; Cd81; CD81 antigen; CD81 antigen (target of antiproliferative antibody 1); CD81 molecule; CVID6; S5.7; Tapa1; Tapa-1; Target of the antiproliferative antibody 1; Tetraspanin28; tetraspanin-28; Tspan28; Tspan-28
Gene Symbols CD81
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 975
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.