Learn More
Invitrogen™ CD81 Monoclonal Antibody (Eat-2 (EAT2)), Functional Grade, eBioscience™

Description
The Eat-2 (EAT2) monoclonal antibody reacts with mouse CD81, a molecule also known as TAPA-1. CD81 is expressed on CD4^+CD8^+ thymocytes and broadly in the periphery with high level expressed by B cells, NK cells, macrophages and dendritic cells and lower level on resting T cells. Activation of T cells induces upregulation of CD81. CD81 is involved in homotypic adhesion of B and T cells as well as costimulation of T cells. The Eat-2 antibody is reported to cross-react with rat CD81. Applications Reported: The Eat-2 (EAT2) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunoblotting (WB). Applications Tested: The Eat-2 (EAT2) antibody has been tested by flow cytometric analysis of mouse thymocytes and splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
Specifications
Specifications
| Antigen | CD81 |
| Applications | Control, Flow Cytometry, Functional Assay, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | Eat-2 (EAT2) |
| Concentration | 1 mg/mL |
| Conjugate | Functional Grade |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene | CD81 |
| Gene Accession No. | P35762 |
| Gene Alias | 26 kDa cell surface protein TAPA-1; CD 81 antigen; Cd81; CD81 antigen; CD81 antigen (target of antiproliferative antibody 1); CD81 molecule; CVID6; S5.7; Tapa1; Tapa-1; Target of the antiproliferative antibody 1; Tetraspanin28; tetraspanin-28; Tspan28; Tspan-28 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.