Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD82 Monoclonal Antibody (C33), PE
GREENER_CHOICE

Catalog No. PIMA528571 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528571 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528571 Supplier Invitrogen™ Supplier No. MA528571
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

This antibody recognizes an extracellular/luminal epitope of CD82, a widely expressed cell surface protein of the tetraspanin family. CD82 is also found in endosome/lysosome compartments. Recommended dilution is for 100 μL of whole blood or 1x10^6 cells in suspension.

This metastasis suppressor gene product is a membrane glycoprotein that is a member of the transmembrane 4 superfamily. Expression of this gene has been shown to be downregulated in tumor progression of human cancers and can be activated by p53 through a consensus binding sequence in the promoter. Its expression and that of p53 are strongly correlated, and the loss of expression of these two proteins is associated with poor survival for prostate cancer patients. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD82
Applications Flow Cytometry
Classification Monoclonal
Clone C33
Concentration 0.025 mg/mL
Conjugate PE
Formulation PBS with 0.2% BSA and 15mM sodium azide; pH 7.4
Gene CD82
Gene Accession No. P27701
Gene Alias 4F9; AA682076; AL023070; C33; C33 antigen; Cd82; CD82 antigen; CD82 antigen (R2 leukocyte antigen antigen detected by monoclonal and antibody IA4)); CD82 antigen (R2 leukocyte antigen, antigen detected by monoclonal and antibody IA4)); CD82 molecule; GR15; IA4; Inducible membrane protein R2; KAI1; Kangai 1 (suppression of tumerigenicity 6 prostate) CD82 antigen; kangai 1 (suppression of tumorigenicity 6), prostate; kangai 1 (suppression of tumorigenicity 6, prostate); Kangai 1 (suppression of tumorigenicity 6, prostate; CD82 antigen (R2 leukocyte antigen, antigen detected by monoclonal and antibody IA4)); metastasis suppressor Kangai-1; Metastasis suppressor Kangai-1 homolog; R2; SAR2; ST6; Suppressor of tumorigenicity 6 protein; Tetraspanin-27; Tspan27; tspan-27
Gene Symbols CD82
Host Species Mouse
Immunogen C91/PL (human HTLV-1+ T cell line).
Purification Method Size-exclusion chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3732
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.