Learn More
Abcam CD9 Exosome Capture Beads, Human, Liquid, 25 Test
Description
- Reactive species: Human
- Form: Liquid
- Storage buffer: 0.09% Sodium azide and 1% Casein
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Format | Beads |
| Buffer | 1% Casein, 0.09% sodium azide |
| Form | Liquid |
| No. of Reactions | 25 Tests |
| Reactivity | Human |
| Target Molecule | CD9 (Exosomes) |
| Cross Reactivity | Human |
| Formulation | Preservative: 0.09% Sodium azide, Constituents: 1% Casein |
| Quantity | 25 Tests |
| For Use With (Application) | Exosome capture |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.