Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD9 Monoclonal Antibody (eBioSN4 (SN4 C3-3A2)), eBioscience™
GREENER_CHOICE

Catalog No. 5012404
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-124-04 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-124-04 Supplier Invitrogen™ Supplier No. 14009882
  • $157.00
    $117.65 / Each of 1

    Save $39.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: The eBioSN4 monoclonal antibody reacts with human CD9. CD9 is a 24 kDa member of the tetraspanin family, whose members are characterized by the presence of 4 hydrophobic transmembrane domains. CD9 is expressed in platelets, eosinophils, basophils, pre-B cells, activated T cells and neural cell lines. Furthermore, CD9 expression has been associated with a malignant phenotype, including expression on 90% of non T cell acute lymphoblastic leukemia cells and on 50% of chronic lymphocytic and acute myeloblastic leukemias. In platelets, CD9 is expressed in alpha-granules and through association with the integrin alphaIIb/betaIII plays a role in platelet aggregation, as demonstrated by the use of anti-human CD9 antibodies. CD9 has also been shown to induce the aggregation of pre-B cell lines, and the adhesion and migration of pre-B cells and Schwann cells. Additionally, it has been demonstrated that CD9 is able to provide a co-stimulatory signal for T cells independently of CD28, in the absence of antigen-presenting cells. Binding of the eBioSN4 monoclonal antibody partially cross-blocks binding of another anti-human CD9 monoclonal antibody, MM2/57. Applications Reported: This eBioSN4 (SN4 C3-3A2) antibody has been reported for use in flow cytometric analysis, immunoprecipitation, and immunohistology staining of frozen tissue sections. Applications Tested: This eBioSN4 (SN4 C3-3A2) antibody has been tested by flow cytometric analysis of normal human platelets. This c...

CD9 antigen is a glycoprotein expressed on the surface of developing B lymphocytes, platelets, monocytes, eosinophils, basophil, stimulated T lymphocytes and by neurons and glial cells in the peripheral nervous system. CD9 belongs to a family of membrane proteins termed tetraspanins which transverse the membrane four times. In pre B cells and platelets, CD9 antigen regulates cell activation and aggregation possibly through an association with the integrin CD41 / CD61 (GPIIb / GPIIIa). CD9 is involved in cell motility, osteoclastogenesis, neurite outgrowth, myotube formation, and sperm-egg fusion, plays roles in cell attachment and proliferation and is necessary for association of heterologous MHC II molecules on the dendritic cell plasma membrane which is important for effective T cell stimulation. CD9 functions in many cellular processes including differentiation, adhesion, and signal transduction, and expression plays a critical role in the suppression of cancer cell motility and metastasis. CD9 is also considered as metastasis suppressor in solid tumors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD9
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Classification Monoclonal
Clone eBioSN4 (SN4 C3-3A2)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CD9
Gene Accession No. P21926
Gene Alias 5H9; 5H9 antigen; antigen CD9; BA2; BA-2/p24 antigen; BTCC-1; CD9; CD9 antigen; CD9 antigen (p24); CD9 molecule; cell growth-inhibiting gene 2 protein; DRAP-27; GIG2; Leukocyte antigen MIC3; MIC3; motility related protein-1; Motility-related protein; MRP-1; p24; sCD 9; sCD9; soluble CD 9; soluble CD9; Tetraspanin29; Tetraspanin-29; transmembrane protein CD9; TSPAN29; Tspan-29
Gene Symbols CD9
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 928
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.