Learn More
Invitrogen™ CD90 Monoclonal Antibody (F15-42-1), FITC

Description
F15-42-1 recognizes the human CD90 cell surface antigen, a ~25 kDa glycoprotein homologous to rat Thy1. The antigen is expressed by a subset of CD34+ve cells in the bone marrow and by prothymocytes within the thymus. CD90 is also expressed extensively within the brain.Mouse anti Human CD90 antibody, clone F15-42-1 is routinely tested in flow cytometry on the MOLT4 cell line.
Specifications
Specifications
| Antigen | CD90 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | F15-42-1 |
| Concentration | 0.1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 0.09% sodium azide |
| Gene | Thy1 |
| Gene Accession No. | P04216 |
| Gene Alias | CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.