Learn More
Invitrogen™ CD90 Monoclonal Antibody (G7), FITC, Invitrogen™
Description
Specifications
Specifications
| Antigen | CD90 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | G7 |
| Concentration | 0.5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with <0.1% sodium azide; pH 7.4 |
| Gene | Thy1 |
| Gene Accession No. | P01831 |
| Gene Alias | CD7; CD90; CDw90; FLJ33325; membrane glycoprotein; T25; Thy 1.2; THY1; Thy-1; Thy-1 antigen; Thy-1 cell surface antigen; thy-1 glycoprotein; thy-1 membrane glycoprotein; Thy-1 protein; Thy-1 T-cell antigen; Thy1.1; Thy1.2; Thy-1.2; thymus cell antigen 1, theta; Thymus cell surface antigen |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.