Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD93 (AA4.1) Monoclonal Antibody (AA4.1), Biotin, eBioscience™, Invitrogen™

Catalog No. 5012009
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-120-09 500 μg
50-120-08 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-120-09 Supplier Invitrogen™ Supplier No. 13589285
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal Antibody

Description: The AA4.1 monoclonal antibody reacts with mouse C1qRp, an approximately 130-140 kDa C-type lectin-like type I transmembrane protein. The AA4.1 antigen was originally identified as an antigen expressed on early stages of B cell development in bone marrow. Adult spleen and fetal liver also have detectable numbers of AA4.1^+ cells. In combination with other markers of hematopoietic progenitor cells such as Thy-1, Sca-1, c-Kit, CD43, and CD24, the bone marrow Lymphoid-Committed Progenitors (CLP) can be segregated into more primitive and more differentiated subsets based on expression of AA4.1. Correlated expression of surface IgM (sIgM), CD23, and AA4.1 antigen has also been used to define three nonproliferative subpopulations of immature/transitional peripheral B cells designated: T1 (AA4.1^+/CD23^-/sIgMhi), T2 (AA4.1^+/CD23^+/sIgMhi), and T3 (AA4.1^+/CD23^+/sIgMlo). AA4.1 is also reported to be expressed in cytoplasmic vesicles in endothelial cells, megakaryoblasts, and platelets. It is reported that monoclonal antibodies AA4.1 and 493 recognize different epitopes of the same molecule. Applications Reported: The AA4.1 antibody has been reported for use in flow cytometric analysis. Applications Tested: The AA4.1 antibody has been tested by flow cytometric analysis of mouse bone marrow and splenocyte cells. This can be used at less than or equal to 0.25 μg per test.

The protein encoded by this gene is a cell-surface glycoprotein and type I membrane protein that was originally identified as a myeloid cell-specific marker. The encoded protein was once thought to be a receptor for C1q, but now is thought to instead be involved in intercellular adhesion and in the clearance of apoptotic cells. The intracellular cytoplasmic tail of this protein has been found to interact with moesin, a protein known to play a role in linking transmembrane proteins to the cytoskeleton and in the remodelling of the cytoskeleton.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD93 (AA4.1)
Applications Flow Cytometry
Classification Monoclonal
Clone AA4.1
Concentration 0.5 mg/mL
Conjugate Biotin
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Cd93
Gene Accession No. O89103
Gene Alias 6030404G09Rik; AA145088; Aa4; AA4.1; AW555904; C1q receptor; C1q receptor 1; C1q/MBL/SPA receptor; C1qR; C1qR(P); C1qr1; C1qRP; Cd93; CD93 antigen; CD93 molecule; CDw93; Cell surface antigen AA4; complement component 1 q subcomponent receptor 1; complement component 1, q subcomponent, receptor 1; complement component C1q receptor; dJ737E23.1; ECSM3; Ly68; ly-68; Lymphocyte antigen 68; matrix-remodeling-associated protein 4; matrix-remodelling associated 4; MXRA4
Gene Symbols Cd93
Host Species Rat
Purification Method Affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17064
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.