Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CD96 Recombinant Protein

Catalog No. 89011793 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-011-793 25 μg
89-011-792 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-011-793 Supplier Abnova™ Supplier No. H00010225P0125
Add to Cart
Add to Cart

Human CD96 full-length ORF recombinant protein with GST-tag at N-terminal

The protein encoded by this gene belongs to the immunoglobulin superfamily. It is a type I membrane protein. The protein may play a role in the adhesive interactions of activated T and NK cells during the late phase of the immune response. It may also function in antigen presentation. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.

  • Theoretical MW (kDa): 69.96
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MEKKWKYCAVYYIIQIHFVKGVWEKTVNTEENVYATLGSDVNLTCQTQTVGFFVQMQWSKVTNKIDLIAVYHPQYGFYCAYGRPCESLVTFTETPENGSRWTLHLRNMSCSVSGRYECMLVLYPEGIQTKIYNLLIQTHVTADEWNSNHTIEIEINQTLEIPCFQNSSSKISSEFTYAWSVEDNGTQETLISQNHLISNSTLLKDRVKLGTDYRLHLSPVQIFDDGRKFSCHIRVGPNKILRSSTTVKVFAKPEIPVIVENNSTDVLVERRFTCLLKNVFPKANITWFIDGSFLHDEKEGIYITNEERKGKDGFLELKSVLTRVHSNKPAQSDNLTIWCMALSPVPGNKVWNISSGKITFLLGSEISSTDPPLSVTESTLDTQPSPASSVSPASKNVFTLSY

Best use within three months from the date of receipt of this protein.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH20749
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 10225
Molecular Weight (g/mol) 69.96
Name CD96 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Source Wheat Germ (in vitro)
Immunogen MEKKWKYCAVYYIIQIHFVKGVWEKTVNTEENVYATLGSDVNLTCQTQTVGFFVQMQWSKVTNKIDLIAVYHPQYGFYCAYGRPCESLVTFTETPENGSRWTLHLRNMSCSVSGRYECMLVLYPEGIQTKIYNLLIQTHVTADEWNSNHTIEIEINQTLEIPCFQNSSSKISSEFTYAWSVEDNGTQETLISQNHLISNSTLLKDRVKLGTDYRLHLSPVQIFDDGRKFSCHIRVGPNKILRSSTTVKVFAKPEIPVIVENNSTDVLVERRFTCLLKNVFPKANITWFIDGSFLHDEKEGIYITNEERKGKDGFLELKSVLTRVHSNKPAQSDNLTIWCMALSPVPGNKVWNISSGKITFLLGSEISSTDPPLSVTESTLDTQPSPASSVSPASKNVFTLSY
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias DKFZp667E2122/MGC22596/TACTILE
Common Name CD96
Gene Symbol CD96
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.