Learn More
Invitrogen™ CD97 Monoclonal Antibody (MEM-180), FITC

Description
This antibody recognizes a unique epitope on CD97, a 75-85 kDa surface glycoprotein of G-protein-coupled receptor family, expressed on activated B and T lymphocytes, monocytes/macrophages, dendritic cells and granulocytes.
Specifications
Specifications
| Antigen | CD97 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MEM-180 |
| Conjugate | FITC |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 |
| Gene | ADGRE5 |
| Gene Accession No. | P48960 |
| Gene Alias | AA409984; Adgre5; adhesion G protein-coupled receptor E5; Cd97; CD97 antigen; CD97 antigen subunit alpha; CD97 antigen subunit beta; CD97 molecule; EGF-TM7 receptor; leukocyte antigen CD97; seven transmembrane helix receptor; seven-span transmembrane protein; seven-transmembrane, heterodimeric receptor associated with inflammation; TM7LN1 |
| Gene Symbols | ADGRE5, CD97 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.