Learn More
Description
This antibody reacts with an extracellular epitope of CD98, a 125 kDa disulfide-linked heterodimer (80 kDa glycosylated heavy chain + 45 kDa non-glycosylated light chain). CD98 is expressed on T lymphocytes (upon activation) and activated NK cells; it is also present at low levels on B lymphocytes, NK cells, monocytes and platelets.
Specifications
Specifications
| Antigen | CD98 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation |
| Classification | Monoclonal |
| Clone | MEM-108 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | SLC3A2 |
| Gene Accession No. | P08195 |
| Gene Alias | 4F2; 4F2 cell-surface antigen heavy chain; 4F2 heavy chain antigen; 4F2hc; 4T2HC; AI314110; antigen defined by monoclonal antibody 4F2, heavy chain; antigen identified by monoclonal antibodies 4F2; antigen identified by monoclonal antibodies 4F2, TRA1.10, TROP4, and T43; Cd98; CD98 antigen; CD98 heavy chain; CD98HC; Ly10; Ly-10; Ly-m10; Lymphocyte activation antigen 4F2 large subunit; MDU1; Mgp-2hc; monoclonal antibody 44D7; NACAE; RGD:3073}; SLC3A2; slc3a2 {ECO:0000312; solute carrier family 3 (activators of dibasic and neutral amino acid transport), member 2; solute carrier family 3 (amino acid transporter heavy chain), member 2; solute carrier family 3 member 2; solute carrier family 3, member 2; type II transmembrane protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
