Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD99 Mouse anti-Human, Clone: 5B6A4, Proteintech
SDP

Catalog No. 501736441 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6441 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6441 Supplier Proteintech Group Inc Supplier No. 603541IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD99 (E2, MIC2) is a transmembrane glycoprotein that is involved in regulation of T cell adhesive properties and programmed cell death distinct from typical apoptosis course. CD99 roles are specific to certain stages of T cell differentiation such as corticothymocytes. CD99R isoform expression is restricted in the hematopoietic system to T, NK and myeloid cells.

Specifications

Antigen CD99
Applications Western Blot
Classification Monoclonal
Clone 5B6A4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CD99
Gene Accession No. P14209
Gene Alias 12E7, CD99, CD99 antigen, CD99 molecule, E2 antigen, MIC2, MIC2X, MIC2Y, Protein MIC2, T cell surface glycoprotein E2
Gene Symbols CD99
Host Species Mouse
Immunogen CD99 Fusion Protein Ag19474
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4267
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.