Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci CD99 Recombinant Protein
SDP

Catalog No. 16594231
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-594-231 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-594-231 Supplier ProSci Supplier No. 914030050
Add to Cart
Add to Cart

Recombinant Protein

This recombinant protein can be used for biological assays. For research use only.

CD99 is a type I transmembrane glycoprotein and the founding member of the CD99 family of molecules. The extracellular domain of CD99 contains no identifiable motifs, its cytoplasmic region, although short, does have signal transduction capability. Cells known to express CD99 include fibroblasts, neutrophils, T cells, double positive thymocytes, CD34+ stem cells, monocytes and endothelial cells. Two types of CD99 isoforms have been classified. Native human CD99 is referred to as the long, or type I isoform. The best studied type II isoform shows an Asp-Gly substitution for the C terminal 27 amino acids. The type I and II isoforms have distinctive signal transduction pathways (FAKsrc for type I PI3K plus srcERK1/2 for type II), and mediate clearly different biological outcomes. Homophilic interaction between CD99 on the neutrophil and CD99 on the endothelial cell regulates the transendothelial migration of neutrophils during inflammation. Human CD99 has 48% aa sequence identity to mouse CD99.

Specifications

Accession Number P14209
Gene ID (Entrez) 4267
Molecular Weight (g/mol) 11.13 kDa
Quantity 0.05 mg
Source Human Cells
Common Name CD99 molecule
Gene Symbol CD99
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag C-6 His tag
Sequence Asp23-Asp122
Content And Storage Lyophilized protein should be stored at -20°C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4°C to 7°C for 2-7 days. Aliquots of reconstituted samples are stable (at -20°C for 3 months.)
Purity or Quality Grade >95% as determined by reducing SDS-PAGE. Endotoxin level less than 0.1 ng/μg (1 IEU/μg) as determined by LAL test.
Synonym CD99 Antigen, 12E7, E2 Antigen, Protein MIC2, T-Cell Surface Glycoprotein E2, CD99, MIC2, MIC2X, MIC2Y
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.