Learn More
Invitrogen™ CD99L2 Polyclonal Antibody

Description
In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CD99-L2 is observed and less than 5% cross-reactivity with recombinant mouse CD99 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | CD99L2 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Cd99l2 |
| Gene Accession No. | Q8BIF0 |
| Gene Alias | AW548191; CD99; Cd99 antigen-like 2; CD99 antigen-like protein 2; CD99 molecule like 2; CD99 molecule-like 2; CD99B; CD99L2; MIC2 (monoclonal Imperial Cancer Research Fund 2)-like 1; MIC2 like 1; MIC2 like-1; MIC2L1; MIC2-like protein 1; protein rhombex-40; Rhombencephalic expression protein 40 kDa; Rhombex40; Rhombex-40; single-pass transmembrane protein; UNQ1964/PRO4486; vms-tm2; Xap89 |
| Gene Symbols | Cd99l2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.