Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc14A Polyclonal Antibody
GREENER_CHOICE

Catalog No. 348100
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-810-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-810-0 Supplier Invitrogen™ Supplier No. 348100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Cdc14A Polyclonal Antibody for Western Blot, ICC/IF, Flow

The protein encoded by this gene is a member of the dual specificity protein tyrosine phosphatase family. It is highly similar to Saccharomyces cerevisiae Cdc14, a protein tyrosine phosphatase involved in the exit of cell mitosis and initiation of DNA replication, suggesting a role in cell cycle control. This protein has been shown to interact with, and dephosphorylate tumor suppressor protein p53, and is thought to regulate the function of p53. Alternative splicing of this gene results in several transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc14A
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene CDC14A
Gene Accession No. Q9UNH5
Gene Alias A830059A17Rik; CDC10 (cell division cycle 10, S. cerevisiae, homolog); cdc14; CDC14 cell division cycle 14 homolog A; CDC14 cell division cycle 14A; CDC14A; CDC14a1; CDC14A2; cell division cycle 14A; DFNB105; Dual specificity protein phosphatase CDC14A; hCDC14
Gene Symbols CDC14A
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of the human Cdc14A protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8556
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.