Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc20 Monoclonal Antibody (BA8)
GREENER_CHOICE

Catalog No. PIMA146040
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA146040 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA146040 Supplier Invitrogen™ Supplier No. MA146040
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cdc20 Monoclonal Antibody for Western Blot, Flow, ELISA

Required for full ubiquitin ligase activity of the anaphase promoting complex/cyclosome (APC/C) and may confer substrate specificity upon the complex. Is regulated by MAD2L1: in metaphase the MAD2L1-CDC20-APC/C ternary complex is inactive and in anaphase the CDC20-APC/C binary complex is active in degrading substrates. The CDC20-APC/C complex positively regulates the formation of synaptic vesicle clustering at active zone to the presynaptic membrane in postmitotic neurons. CDC20-APC/C-induced degradation of NEUROD2 induces presynaptic differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc20
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone BA8
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene cdc20.S
Gene Accession No. 0
Gene Alias 2310042N09Rik; bA276H19.3; C87100; cdc20; CDC20 cell division cycle 20 homolog; cdc20.S; CDC20A; cell cycle protein p55CDC; cell division cycle 20; cell division cycle 20 homolog; cell division cycle 20 S homeolog; cell division cycle protein 20 homolog; fizzy1; mmCdc20; p55cdc; XELAEV_18025163mg; X-FZY
Gene Symbols cdc20.S
Host Species Mouse
Immunogen Peptide from the carboxy terminus of the protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 379130
Target Species Frog
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.