Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ CDC23 Recombinant Protein

Catalog No. 89968705 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-968-705 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-968-705 Supplier Abnova™ Supplier No. H00008697P01
Add to Cart
Add to Cart

Human CDC23 full-length ORF recombinant protein with GST-tag at N-terminal

The protein encoded by this gene shares strong similarity with Saccharomyces cerevisiae Cdc23, a protein essential for cell cycle progression through the G2/M transition. This protein is a component of anaphase-promoting complex (APC), which is composed of eight protein subunits and highly conserved in eukaryotic cells. APC catalyzes the formation of cyclin B-ubiquitin conjugate that is responsible for the ubiquitin-mediated proteolysis of B-type cyclins. This protein and 3 other members of the APC complex contain the TPR (tetratricopeptide repeat), a protein domain important for protein-protein interaction.

  • Molecular Weight: 90.75kDa
  • Preparation Method: in vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 Fast Flow
  • Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH 8.0 in the elution buffer

Sequence: MVPVAVTAAVAPVLSINSDFSDLREIKKQLLLIAGLTRERGLLHSSKWSAELAFSLPALPLAELQPPPPITEEDAQDMDAYTLAKAYFDVKEYDRAAHFLHGCNSKKAYFLYMYSRYLSGEKKKDDETVDSLGPLEKGQVKNEALRELRVELSKKHQARELDGFGLYLYGVVLRKLDLVKEAIDVFVEATHVL
PLHWGAWLELCNLITDKEMLKFLSLPDTWMKEFFLAHIYTELQLIEEALQKYQNLIDVGFSKSSYIVSQIAVAYHNIRDIDKALSIFNELRKQDPYRIENMDTFSNLLYVRSMKSELSYLAHNLCEIDKYRVETCCVIGNYYSLRSQHEKAALYFQRALKLNPRYLGAWTLMGHEYMEMKNTSAAIQAYRHAIEVNKRDYRAWYGLGQTYEILKMPFYCLYYYRRAHQLRPNDSRMLVALGECYEKLNQLVEAKKCYWRAYAVGDVEKMALVKLAKLHEQLTESEQAAQCYIKYIQDIYSCGEIVEHLEESTAFRYLAQYYFKCKLWDEASTCAQKCCAFNDTREEGKALLRQILQLRNQGETPTTEVPAPFFLPASLSANNTPTRRVSPLNLSSVTP

Best when used within three months from the date of receipt.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Accession Number AAH17713
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50 mM Tris HCl, 10 mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 8697
Molecular Weight (g/mol) 90.75 kDa
Name CDC23 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue
Quantity 2 μg
Source Wheat Germ (in vitro)
Immunogen MVPVAVTAAVAPVLSINSDFSDLREIKKQLLLIAGLTRERGLLHSSKWSAELAFSLPALPLAELQPPPPITEEDAQDMDAYTLAKAYFDVKEYDRAAHFLHGCNSKKAYFLYMYSRYLSGEKKKDDETVDSLGPLEKGQVKNEALRELRVELSKKHQARELDGFGLYLYGVVLRKLDLVKEAIDVFVEATHVLPLHWGAWLELCNLITDKEMLKFLSLPDTWMKEFFLAHIYTELQLIEEALQKYQNLIDVGFSKSSYIVSQIAVAYHNIRDIDKALSIFNELRKQDPYRIENMDTFSNLLYVRSMKSELSYLAHNLCEIDKYRVETCCVIGNYYSLRSQHEKAALYFQRALKLNPRYLGAWTLMGHEYMEMKNTSAAIQAYRHAIEVNKRDYRAWYGLGQTYEILKMPFYCLYYYRRAHQLRPNDSRMLVALGECYEKLNQLVEAKKCYWRAYAVGDVEKMALVKLAKLHEQLTESEQAAQCYIKYIQDIYSCGEIVEHLEESTAFRYLAQYYFKCKLWDEASTCAQKCCAFNDTREEGKALLRQILQLRNQGETPTTEVPAPFFLPASLSANNTPTRRVSPLNLSSVTP
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias APC8
Common Name CDC23
Gene Symbol CDC23
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.