Learn More
Diagnostic Biosystems CDC25A CDC25A
Supplier:  Diagnostic Biosystems MOB337
Encompass_Preferred
This antibody reacts with a 65 kDa protein known as Cdc25A. The activity of cyclin-dependent kinases in cell cycle is regulated by the phosphorylation status, which is controlled by the antagonistic action of wee 1 kinase and Cdc25 phosphatases. Three CDC25 genes are present in human cells: CDC25A, CDC25B and CDC25C. Cdc25A and Cdc25B are expressed throughout the cell cycle, with peak expression in G1 for Cdc25A and in both G1-S phase and G2 for Cdc25B. Cdc25C is mainly expressed in G2 phase.\n
Catalog No. 50-238-5657
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.