Learn More
Invitrogen™ Cdc25A Monoclonal Antibody (DCS121)

Description
Cdc25A Monoclonal Antibody for Western Blot, IP
Specifications
Specifications
| Antigen | Cdc25A |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | DCS121 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | CDC25A |
| Gene Accession No. | P30304 |
| Gene Alias | CDC25 isoform A1-CAG; CDC25A; CDC25A2; CDC25A2-CAG isoform; Cdc25m3; cell division cycle 25A; D9Ertd393e; dual specificity phosphatase Cdc25A; EC 3.1.3.48; M-phase inducer phosphatase 1; MPIP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.