Learn More
TARGETMOL CHEMICALS INC CDC25B-IN-1 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T10723
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CDC25B-IN-1 is an inhibitor of CDC25B with a Ki of 8.5 uM. CDC25B-IN-1 increases the G2/M phase by inhibiting cell proliferation and colony formation. purity: 97%
Catalog No. 50-258-1684
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.