Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC2L5 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501560445
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0445 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0445 Supplier Bethyl Laboratories, Inc Supplier No. A301458A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Bovine

The protein encoded by this gene is a member of the cyclin-dependent serine/threonine protein kinase family. Members of this family are well known for their essential roles as master switches in cell cycle control. Some of the cell cycle control kinases are able to phosphorylate proteins that are important for cell differentiation and apoptosis, thus provide connections between cell proliferation, differentiation, and apoptosis. Proteins of this family may also be involved in non-cell cycle-related functions, such as neurocytoskeleton dynamics. The exact function of this protein has not yet been determined. It has unusually large N- and C-termini and is ubiquitously expressed in many tissues. Two alternatively spliced variants are described.

Specifications

Antigen CDC2L5
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CDK13
Gene Accession No. Q14004
Gene Alias CDC2L, CDC2L5, CHED, hCDK13, CDC2-related protein kinase 5, cell division cycle 2-like protein kinase 5, cell division protein kinase 13, cholinesterase-related cell division controller
Gene Symbols CDK13
Host Species Rabbit
Immunogen Between 1125 and 1175
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8621
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.