Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CDC42BPA Monoclonal Antibody (OTI7B9), TrueMAB™, OriGene
SDP

Catalog No. 501681979
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1979 Supplier OriGene Supplier No. TA808223
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MRCK alpha (myotonic dystrophy kinase-related Cdc42-binding kinase alpha) is a member of the dystrophia myotonica protein kinase (DMPK) family that functions downstream of Cdc42 in actin cytoskeletal reorganization. It is a serine/threonine kinase with multiple functional domains, including phorbol ester-responsive C1 domain. Three independent coiled-coil domains and the N-terminal region preceding the kinase domain are responsible for intermolecular interactions leading to MRCK alpha multimerization. The transautophosphorylation process is critical for regulation of MRCK alpha catalytic activities. Binding of phorbol esters to MRCK releases its autoinhibition, allowing N-terminal dimerization and subsequent kinase activation.

Specifications

Antigen CDC42BPA
Applications Western Blot
Classification Monoclonal
Clone OTI7B9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CDC42BPA
Gene Accession No. Q5VT25
Gene Alias MRCK, MRCKA, PK428
Gene Symbols CDC42BPA
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1419-1638 of human CDC42BPA produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8476
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.