Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cdc42EP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103722
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103722 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103722 Supplier Invitrogen™ Supplier No. PA5103722
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total BORG5.

CDC42 is a member of the Rho GTPase family that regulates multiple cellular activities, including actin polymerization. The protein encoded by this gene is a CDC42 binding protein that mediates actin cytoskeleton reorganization at the plasma membrane. This protein is secreted and is primarily found in bone marrow.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cdc42EP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CDC42EP1
Gene Accession No. Q00587, Q91W92
Gene Alias 1810058K22Rik; 55 kDa bone marrow stromal/endothelial cell protein; AA980734; binder of Rho GTPase 5; binder of Rho GTPases 5; Borg5; CDC42 effector protein (Rho GTPase binding) 1; cdc42 effector protein 1; Cdc42ep1; cdc42ep1 {ECO:0000312; CEP1; EMBL:AAI28745.1}; MSE55; serum constituent protein; serum protein MSE55
Gene Symbols CDC42EP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CDC42EP1(Accession Q00587), corresponding to amino acid residues R53-P103.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104445, 11135
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.