Learn More
Invitrogen™ Cdc42EP2 Polyclonal Antibody

Description
Recommended positive controls: mouse heart. Predicted reactivity: Mouse (89%), Rat (87%), Rhesus Monkey (99%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Cdc42EP2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | CDC42EP2 |
| Gene Accession No. | O14613, Q8JZX9 |
| Gene Alias | 1110008C05Rik; binder of Rho GTPases 1; Borg1; CDC42 effector protein (Rho GTPase binding) 2; CDC42 effector protein 2; CDC42EP2; CEP2; CRIB-containing BOGR1 protein; D19Bwg1013e |
| Gene Symbols | CDC42EP2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.