Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CDC47/MCM7 Ab-2, Mouse Monoclonal Antibody

Catalog No. MS862R1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MS862R1 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS862R1 Supplier Epredia™ Supplier No. MS862R1

Obtain accurate, reproducible results in immunohistochemistry, western blotting, immunofluorescence and immunoprecipitation experiments.

hCDC47 is a human member of the MCM family, which allows DNA to replicate once per cell cycle. In quiescent cells, human MCM7 (hMCM7) mRNA is almost undetectable. Stimulation of cells to enter the cell cycle results in induction of hMCM7 expression. The hCDC47 protein expression and localization is found in nuclei of the proliferative components of normal lymph nodes, bone marrow, epidermis and mucosa. Malignant tumors from several organs contained more nuclear hCDC47-positive cells than their normal counterparts. These results indicate that hCDC47 may play a role in normal and neoplastic cell growth in vivo.

Host Species: Mouse

Clone: 47DC141; same as DCS-141

Isotype: IgG1

Species Reactivity: Human, Mouse, Rat, Dog and Xenopus. Others not known.

Epitope: Not determined

Immunogen: Recombinant hCDC47 protein

Molecular Weight: 80 kDa

Positive Control: MAD109, or PC12 cells, Tonsil or breast cancer

Cellular Localization: Nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CDC47/MCM7 Ab-2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 47DC141; same as DCS-141
Conjugate Unconjugated
Formulation Ready-to-use for IHC (UVLP)
Host Species Mouse
Immunogen Recombinant hCDC47 protein
Quantity 1 mL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat, Xenopus species
Form Prediluted
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.