Learn More
Description
It is not recommended to aliquot this antibody.
Specifications
Specifications
| Antigen | CDCP1 |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 66 μg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.01M HEPES with 50% glycerol, 0.15M NaCl, 100μg/mL BSA and no preservative; pH 7.5 |
| Gene | Cdcp1 |
| Gene Accession No. | Q9H5V8 |
| Gene Alias | 9030022E12Rik; AA409659; CD318; CDCP1; CUB domain containing protein 1; CUB domain-containing protein 1; E030027H19Rik; gp140; Membrane glycoprotein gp140; RGD1305578; SIMA135; Subtractive immunization M plus HEp3-associated 135 kDa protein; transmembrane and associated with src kinases; TRASK; UNQ2486/PRO5773 |
| Gene Symbols | Cdcp1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
