Learn More
Invitrogen™ CDH17 Recombinant Human Monoclonal Antibody (10B3)

Description
CDH17 Recombinant Monoclonal Antibody for Flow, ELISA
Specifications
Specifications
| Antigen | CDH17 |
| Applications | ELISA, Flow Cytometry, Neutralization |
| Classification | Recombinant Monoclonal |
| Clone | 10B3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| Gene | Cdh17 |
| Gene Accession No. | Q12864 |
| Gene Alias | BILL-cadherin; Cadherin; cadherin 17; cadherin 17, LI cadherin (liver-intestine); cadherin-16; cadherin-17; CDH16; CDH17; FLJ26931; HPT1; HPT-1; HPT-1 cadherin; HPT-1/LI; human intestinal peptide-associated transporter HPT-1; human peptide transporter 1; intestinal peptide-associated transporter HPT-1; LI cadherin; LI-cadherin; Liver-intestine cadherin; MGC138218; MGC142024; P130 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.