Learn More
Selleck Chemical LLC Roscovitine (Seliciclib) 200mg 186692-46-6 CYC202
SDP
Supplier:  Selleck Chemical LLC S1153200MG
Encompass_Preferred
Roscovitine (Seliciclib, CYC202) is a potent and selective CDK inhibitor for Cdc2, CDK2 and CDK5 with IC50 of 0.65 ?M, 0.7 ?M and 0.16 ?M in cell-free assays. It shows little effect on CDK4/6. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-836-3
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.