Learn More
Enzo Life Sciences CDK1 cdc 2 monoclonal antibody (5F6) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCC101E
Encompass_Preferred
Clone: 5F6. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of human Cdk1 (cdc2). UniProt ID: P06493. Species reactivity: Human, Mouse, Rat, Rabbit. Applications: IHC, IP, WB. Recommended Dilutions: Immunohistochemistry (5µg/ml) Immunoprecipitation (2µg/200µl) Western Blot (1-5µg/ml, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-200-7237
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.