Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CDK11B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588789
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588789 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588789 Supplier Invitrogen™ Supplier No. PA588789
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGDEKDSWKV KTLDEILQEK KRRKEQEEKA EIKRLKNSDD RDSKRDSLEE GELRDHRMEI TIRNSPYRRE DSMEDRGEED DSLAIKPPQQ MSRKEKAHHR KDEKRKEKRR HRSHSAEGGK HARVKEKERE; Positive Samples: HeLa, Mouse brain; Cellular Location: Cytoplasm, Nucleus.

The PITSLRE beta1 protein, a distantly related member of the Cdk family of protein kinases, induces apoptosis after low levels of ectopic expression. Apoptosis, or programmed cell death, is similarly induced by ectopic expression of an amino terminal deletion mutant retaining the catalytic and carboxyterminal domains of PITSLRE beta1, but not by other mutants lacking Histone H1 kinase activity or by other Cdk family members. The terminology for the ten isoforms of the PITSLRE subfamily of proteins is based on the conserved PSTAIRE box region of Cdc2 p34. Depending on which of the PITSLRE genes produce the protein, the cDNA and protein are designated alpha, beta or gamma (i.e., PITSLRE A gene, alpha; PITSLRE B gene, beta and PITSLRE C gene, gamma). Some of the isoforms such as PITSLRE alpha1 (T cells) and PITSLRE beta1 (B cells and brain), are expressed in specific cell types, while others are expressed ubiquitously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CDK11B
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CDK11B
Gene Accession No. P21127, P24788
Gene Alias AA989746; Cdc11b; CDC2L1; Cdc2l2; CDC2L3; CDC-related protein kinase p58; CDK11; CDK11 p110; CDK11 p46; CDK11 p58; CDK11A; Cdk11b; CDK11-p110; CDK11-p46; CDK11-p58; cell division cycle 2 homolog-like 1; cell division cycle 2 homolog-like 2; cell division cycle 2-like 1; cell division cycle 2-like 1 (PITSLRE proteins); cell division cycle 2-like 2; cell division cycle 2-like protein kinase 1; Cell division protein kinase 11; cell division protein kinase 11B; CLK-1; cyclin dependent kinase 11B; cyclin-dependent kinase 11; Cyclin-dependent kinase 11B; galactosyltransferase-associated protein kinase p58/GTA; p58; p58 CLK-1; p58CDC2L1; p58CLK-1; PITSLRE proteins; PITSLRE serine/threonine-protein kinase CDC2L1; PITSLREA; PK58
Gene Symbols CDK11B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human CDK11B (NP_2770212).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12537, 984
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.