Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC CDK12-IN-E9 10mg | 2020052-55-3 | 434.53 | C24H30N6O2 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY117203A10MG

Encompass_Preferred

CDK12-IN-E9 is a research-grade small-molecule inhibitor that selectively and covalently inhibits cyclin-dependent kinase 12 (CDK12) and non-covalently inhibits CDK9. It is supplied as a solid for use in cellular and biochemical studies of transcriptional regulation and DNA damage response.

  • Potent covalent CDK12 inhibition (IC50 23.9 nM).
  • Non-covalent CDK9 activity (IC50 932 nM).
  • High reported purity (~99.7%).
  • Molecular weight 434.53 g/mol; formula C24H30N6O2.
  • Soluble in DMSO (125 mg/mL with ultrasonic).
  • Available in small research pack sizes suitable for early-stage experiments.

Catalog No. 50-002-27800


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.